missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A66
GREENER_CHOICE

Product Code. 18798589
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18798589

Brand: Tocris Bioscience™ 5595/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PI 3-kinase p110α inhibitor

A66 is a potent and selective PI 3-kinase p110α inhibitor (IC50 = 32 nM). Exhibits >100-fold selectivity for p110α over other PI 3-kinase isoforms. Inhibits Akt signaling and tumor growth in SK-OV-3 xenografts in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-N1-[2-(1,1-Dimethylethyl)-4′-methyl[4,5′-bithiazol]-2′-yl]-1,2-pyrrolidinedicarboxamide
CAS 1166227-08-2
Quantity 10 mg
Target PI 3-Kinase Inhibitors
Molecular Formula C17H23N5O2S2
Formula Weight 393.53
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.