missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAAS Antibody (OTI5B10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71358V
This item is not returnable.
View return policy
Description
AAAS Monoclonal specifically detects AAAS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| AAAS | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AAAS | |
| Recombinant protein fragment corresponding to amino acids 322-546 of human AAAS (NP_056480) produced in E.coli. | |
| 0.1 mL | |
| 8086.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5B10 | |
| Western Blot, Immunohistochemistry | |
| AAA, AAASb, achalasia, adrenocortical insufficiency, alacrimia, achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A), ADRACALA, Adracalin, ALADIN, Allgrove, triple-A, DKFZp586G1624 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction