missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAAS Monoclonal Antibody (OTI7H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808617
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the WD-repeat family of regulatory proteins and may be involved in normal development of the peripheral and central nervous system. The encoded protein is part of the nuclear pore complex and is anchored there by NDC1. Defects in this gene are a cause of achalasia-addisonianism-alacrima syndrome (AAAS), also called triple-A syndrome or Allgrove syndrome. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| AAAS | |
| Monoclonal | |
| Unconjugated | |
| AAAS | |
| AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8086 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7H7 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NRG9 | |
| AAAS | |
| Human recombit protein fragment corresponding to amino acids 322-546 of human AAAS produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction