missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAAS Monoclonal Antibody (OTI7H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808617S
This item is not returnable.
View return policy
Description
AAAS Monoclonal antibody specifically detects AAAS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| AAAS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NRH1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57002 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7H7 | |
| Unconjugated | |
| YAE1D1 | |
| AAA; Aaas; AAASb; achalasia, adrenocortical insufficiency, alacrimia; achalasia, adrenocortical insufficiency, alacrimia (Allgrove, triple-A); ADRACALA; Adracalin; ALADIN; aladin WD repeat nucleoporin; Allgrove, triple-A; D030041N15Rik; GL003 | |
| Full length human recombinant protein of human YAE1D1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?