Learn More
Carbolite™ AAF 11/7 Chamber Furnace
Click to view available options
Dimensions (D x W x H) Interior:
3.5 x 6.75 x 18in.
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 7 L |
| Temperature (Metric) | 1100°C |
| Type | Chamber Furnace |
| Includes | Programmable Temperature Controller |
| Dimensions (D x W x H) Interior | 3.5 x 6.75 x 18in. |
| Width (Metric) | 431.8 mm |
| Width (Metric) Interior | 171.45 mm |
| Height (Metric) | 736.6 mm |
| Height (Metric) Interior | 88.9 mm |
| Depth (Metric) | 647.7 mm |
| Show More |