Learn More
AAMDC Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97175APC
Product Code. 18617752
  • € 407.00 / 0.10mL
Description
AAMDC Polyclonal antibody specifically detects AAMDC in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
APC
Immunohistochemistry-Paraffin
Rabbit
Produced in rabbits immunized with E. coli-derived Human AAMDC fragment.