missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAMDC Antibody (OTI1C4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46589
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
AAMDC Monoclonal specifically detects AAMDC in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| C11orf67 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| adipogenesis associated, Mth938 domain containing, C11orf67, chromosome 11 open reading frame 67, CK067, FLJ21035, hypothetical protein LOC28971, MGC3367, PTD015 | |
| Mouse | |
| 13.2 kDa | |
| 0.1 mL | |
| metabolism | |
| 28971 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI1C4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| AAMDC | |
| Full length human recombinant protein of human C11orf67(NP_078960) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction