missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAMDC Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97175PE
This item is not returnable.
View return policy
Description
AAMDC Polyclonal antibody specifically detects AAMDC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PE | |
| Antigen and protein A Affinity-purified | |
| 28971 | |
| Purified |
| PBS | |
| 0.1 mL | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction