Learn More
Sino Biological™ AARS Recombinant Mouse Protein, His tag
Expressed from DNA sequence encoding mouse AARS (Met 1-Asn 968) fused to polyhistidine tag at C-terminus
Merk: Sino Biological™ 50384M08B50
Overige gegevens : Gewicht : 0.01000kg
Beschrijving
- Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
Specificatie
Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications | |
234734 | |
≥85% by SDS-PAGE | |
Mouse | |
<1 EU/μg | |
AARS | |
Other Proteins | |
AARS Recombinant Mouse Protein |
Lyophilized | |
108.3kd | |
50μg | |
Full Length, Recombinant | |
Alanyl-Trna Synthetasep | |
Assay Standard | |
Insect | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.