missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARSD1 Antibody (Tsp232A), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12887IR
This item is not returnable.
View return policy
Description
AARSD1 Monoclonal specifically detects AARSD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AARSD1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AARSD1 | |
| Recombinant tsp23 | |
| 0.1 mL | |
| 80755 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Tsp232A | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 19kDa. Detects Tsp23, no cross-reactivity to p23. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction