missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARSD1 Antibody (Tsp232A), Novus Biologicals™
Mouse Monoclonal Antibody
€ 186.00
Specifications
| Antigen | AARSD1 |
|---|---|
| Clone | Tsp232A |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18743284
|
Novus Biologicals
NBP2-12887-0.025mg |
0.025 mg | |||||||
Description
AARSD1 Monoclonal specifically detects AARSD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Microarray.Specifications
| AARSD1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| 80755 | |
| Recombinant tsp23 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Tsp232A | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| alanyl-tRNA editing protein Aarsd1, alanyl-tRNA synthetase domain containing 1, Alanyl-tRNA synthetase domain-containing protein 1, MGC2744 | |
| AARSD1 | |
| IgG1 κ | |
| Protein G purified | |
| Detects approx 19kDa. Detects Tsp23, no cross-reactivity to p23. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title