missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: TSGIFTDTSS DGLQARRPDV VPAFRSLQKQ VGTPDSLDSL DIPSSASDGG YEVFSPSATG PSGGQPRALD SGYDTENYES PEFVLKEAQE GCEPQAFAEL ASEGEGPGPE TRLSTSLSGL NEKNPYRDSA Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
The protein encoded by this gene contains a tyrosine kinase domain at the N-terminus and a proline-rich domain at the C-terminus. This gene is induced during apoptosis, and expression of this gene may be a necessary pre-requisite for the induction of growth arrest and/or apoptosis of myeloid precursor cells. This gene has been shown to produce neuronal differentiation in a neuroblastoma cell line.
Specifications
Specifications
| Antigen | AATK |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AATK |
| Gene Accession No. | Q6ZMQ8 |
| Gene Alias | Aatk; AATYK; aatyk1; apoptosis associated tyrosine kinase; apoptosis-associated tyrosine kinase; apoptosis-associated tyrosine kinase 1B; brain apoptosis-associated tyrosine kinase; CDK5-binding protein; KIAA0641; Lemur tyrosine kinase 1; LMR1; LMTK1; mKIAA0641; p35-binding protein; p35BP; PPP1R77; protein phosphatase 1, regulatory subunit 77; serine/threonine-protein kinase LMTK1 |
| Gene Symbols | AATK |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
