missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AATK/Serine/threonine-protein kinase LMTK1 Antibody (456006), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4866G
This item is not returnable.
View return policy
Description
AATK/Serine/threonine-protein kinase LMTK1 Monoclonal antibody specifically detects AATK/Serine/threonine-protein kinase LMTK1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| AATK/Serine/threonine-protein kinase LMTK1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 456006 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| AATYKp35-binding protein, apoptosis-associated tyrosine kinaseCDK5-binding protein, Brain apoptosis-associated tyrosine kinase, EC 2.7.11, KIAA0641p35BP, Lemur tyrosine kinase 1, LMR1EC 2.7.11.1, LMTK1AATYK1, serine/threonine-protein kinase LMTK1 | |
| E. coli-derived recombinant human AATK, Asn421-Ala589, Accession # AAH47378 | |
| 0.1 mL | |
| Protein Kinase | |
| 9625 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur