missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AAV4 Virus Polyclonal Antibody

Description
PA1-23358 detects Adeno-Associated 4 Virus in Virus samples.
Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease. The virus causes a very mild immune response, lending further support to its apparent lack of pathogenicity.
Specifications
Specifications
| Antigen | AAV4 Virus |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | AAV; AAV-4; AdV4; AV4; HADV-4; Lentivirus |
| Host Species | Rabbit |
| Immunogen | Anti-Adeno-Associated Virus (AAV) polyclonal antiserum. |
| Purification Method | Affinity Chromatography |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?