missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABAT Monoclonal Antibody (OTI7C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806964
This item is not returnable.
View return policy
Description
ABAT (4-aminobutyrate aminotransferase) is responsible for catabolism of gamma-aminobutyric acid (GABA), an important, mostly inhibitory neurotransmitter in the central nervous system, into succinic semialdehyde. The active enzyme is a homodimer of 50-kD subunits complexed to pyridoxal-5-phosphate. The protein sequence is over 95% similar to the pig protein. GABA is estimated to be present in nearly one-third of human synapses. ABAT in liver and brain is controlled by 2 codomit alleles with a frequency in a Caucasian population of 0.56 and 0.44. The ABAT deficiency phenotype includes psychomotor retardation, hypotonia, hyperreflexia, lethargy, refractory seizures, and EEG abnormalities. Multiple alternatively spliced transcript variants encoding the same protein isoform have been found for this gene.Specifications
| ABAT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P80404 | |
| ABAT | |
| Human recombit protein fragment corresponding to amino acids 29-323 of human ABAT produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI7C4 | |
| Unconjugated | |
| ABAT | |
| GABA-AT, GABAT, NPD009 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 18 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction