Learn More
ABCA1 Antibody (5A1-1422.11) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NB400-165JF635
Product Code. 30171199
  • € 574.00 / 0.10mL
Description
ABCA1 Monoclonal antibody specifically detects ABCA1 in Mouse samples. It is validated for Flow Cytometry,Western Blot (Negative)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ABCA1
Monoclonal
Janelia Fluor 635
ABC-1, ABC1ATP-binding cassette transporter A1, ATP-binding cassette 1, ATP-binding cassette transporter 1, ATP-binding cassette, sub-family A (ABC1), member 1, CERP, Cholesterol efflux regulatory protein, EC 2.7.7.8, EC 2.8.1.8, EC 3.6.3, EC 3.6.3.41, FLJ14958, HDLDT1, membrane-bound, MGC164864, MGC165011, TGDATP-binding cassette sub-family A member 1
Mouse ABCA1 Antibody (5A1-1422.11) expressing HeLa stable transfectants. [UniProt# P41233]
0.1 mL
ABC Transporters, Astrocyte Markers, Cancer, Cardiovascular Biology, Cellular Markers, Cholesterol Metabolism, Diabetes Research, Glia Markers, Golgi Apparatus Markers, Lipid and Metabolism, Neuroscience, Signal Transduction
19
Store at 4°C in the dark.
IgG2a κ
Flow Cytometry, Western Blot (Negative)
5A1-1422.11
50mM Sodium Borate
Rat
Protein A or G purified