missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCA2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17857922
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17857922 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17857922 Supplier Invitrogen™ Supplier No. PA5142377

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_001597.2; NP_997698.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.

Probable transporter, its natural substrate has not been found yet. May have a role in macrophage lipid metabolism and neural development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Abca2
Gene Accession No. Q9BZC7
Gene Alias Abc2; ABCA2; abca2 {ECO:0000312; AI413825; ATP binding cassette subfamily A member 2; ATP-binding cassette 2; ATP-binding cassette sub-family A member 2; ATP-binding cassette transporter 2; ATP-binding cassette, subfamily A (ABC1), member 2; ATP-binding cassette, sub-family A (ABC1), member 2; ATP-binding cassette, sub-family A, member 2; D2H0S1474E; KIAA1062; mKIAA1062; RGD:620238}
Gene Symbols Abca2
Host Species Goat
Immunogen Peptide with sequence C-KKQSDNLEQQETEP.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.