missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCA8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17772144
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17772144 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17772144 Supplier Invitrogen™ Supplier No. PA5120749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, HepG2, U-251MG. Immunogen sequence: NEYGHRRPPL FFLKSSFWSQ TQKTDHVALE DEMDADPSFH DSFEQAPPEF QGKEAIRIRN VTKEYKGKPD K.

ATP-binding cassette (ABC) transporters are an evolutionarily conserved family of proteins that catalyze the transport of molecules across extracellular and intracellular membranes by harnessing the energy of ATP hydrolysis. ABCA8 (ATP-binding cassette, sub-family A (ABC1), member 8) is a 1, 581 amino acid multi-pass membrane protein that contains two ABC transporter domains and belongs to the ABC transporter family. Expressed in a variety of tissues with particularly high expression in liver, heart and skeletal muscle, ABCA8 functions as an ATP-dependent lipophilic drug transporter that exists as multiple alternatively spliced isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene ABCA8
Gene Accession No. O94911
Gene Alias Abca8; Abca8a; Abca8b; ATP binding cassette subfamily A member 8; ATP-binding cassette sub-family A member 8; ATP-binding cassette sub-family A member 8-A; ATP-binding cassette sub-family A member 8-B; ATP-binding cassette, subfamily A (ABC1), member 8; ATP-binding cassette, sub-family A (ABC1), member 8; ATP-binding cassette, subfamily A (ABC1), member 8a; ATP-binding cassette, sub-family A (ABC1), member 8a; ATP-binding cassette, sub-family A (ABC1), member 8b; KIAA0822
Gene Symbols ABCA8, Abca8a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 425-495 of human ABCA8 (NP_009099.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10351, 303637, 303638
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.