missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCA9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15720812
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15720812 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15720812 Supplier Invitrogen™ Supplier No. PA563027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FWFQHGRANH VVLENETDSD PTPNDCFEP Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.

ABCA9 (ATP-binding cassette, sub-family A (ABC1), member 9) is a 1,624 amino acid protein belonging to the ABC transporter superfamily and the ABCA family. The ABC1 subfamily is the only major ABC subfamily exclusive to multicellular eukaryotes. Ubiquitously expressed, with high expression in heart, brain and fetal tissues, ABCA9 is a multi-pass membrane protein that contains two ABC transporter domains and exists as five alternatively spliced isoforms. ABCA9 exhibits membrane subcellular localization and may play a role in monocyte differentiation and lipid homeostasis. ABCA9 is up-regulated during monocyte differentiation into macrophages and down-regulated by cholesterol loading of macrophages. Spanning 85 kb, ABCA9 contains 39 exons, with a non-coding first exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCA9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABCA9
Gene Accession No. Q8IUA7
Gene Alias ABCA9; ATP binding cassette subfamily A member 9; ATP-binding cassette A9; ATP-binding cassette sub-family A member 9; ATP-binding cassette transporter sub-family A member 9; ATP-binding cassette, subfamily A (ABC1), member 9; ATP-binding cassette, sub-family A (ABC1), member 9; D630040K07Rik; DKFZp686F2450; EST640918; MGC75415
Gene Symbols ABCA9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABCA9. Recombinant protein control fragment (Product #RP-101389).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10350
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.