Learn More
Abcam Human/Mouse/Rat beta Amyloid 1-42 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Product Code. 32111757 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32111757 10 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32111757 Supplier Abcam Supplier No. ab25358310x96Tests
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Antibody, suitable for sELISA.
  • Applications: sELISA
  • Species Reactivity: Mouse, Rat and Human
Specifications
Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols APP
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Mouse, Rat and Human.
Target Species Mouse, Rat, Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Ms, Rt, Hu
Validated Application abrv sELISA