missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB1 Monoclonal Antibody (OTI10H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806730S
This item is not returnable.
View return policy
Description
ABCB1 Monoclonal antibody specifically detects ABCB1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ABCB1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z3V4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 89910 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| OTI10H3 | |
| Unconjugated | |
| UBE3B | |
| ABC superfamily; P-glycoprotein; ABC20; ABCB1; Abcb1a; Abcb1b; Abcb4; ATP binding cassette subfamily B member 1; ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette sub-family B member 1; ATP-binding cassette sub-family B member 1A; ATP-binding cassette sub-family B member 1B; ATP-binding cassette transporter subfamily B, member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette, subfamily B (MDR/TAP), member 1A; ATP-binding cassette, sub-family B (MDR/TAP), member 1A; ATP-binding cassette, subfamily B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, subfamily B, member 1A; ATP-binding cassette, subfamily B, member 1B; ATP-dependent translocase ABCB1; ATP-dependent translocase ABCB1; multidrug r | |
| Human recombinant protein fragment corresponding to amino acids 768-1068 of human UBE3B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction