missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCB11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103659
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ABCB11.
ABCB11 is involved in the ATP-dependent secretion of bile salts into the canaliculus of hepatocytes. It is expressed predominatly, if not exclusively, in the liver, where it is further localized to the canilicular microvilli and to subcanilicular vesicles fo the hepatocytes. Structurally, ABCB11 is a multifunctional polypeptide with two homologus halves, each containing a hydrophobic membrane-anchoring domain and an ATP binding cassette (ABC) domain. Defects in ABCB11 are the cause of progressive familial intrahepatic cholestasis 2 (PFIC2). PFIC2 is an inherited liver disease of childhood which is characterized by cholestasis and normal serum gamma-glutamyltransferase activity. Defects in ABCB11 are also found in cases of chronic intrahepatic cholestasis without obvious familial history of chronic liver disease.
Specifications
| ABCB11 | |
| Polyclonal | |
| Unconjugated | |
| ABCB11 | |
| ABC member 16, MDR/TAP subfamily; ABC16; Abcb11; ATP binding cassette subfamily B member 11; ATP-binding cassette sub-family B member 11; ATP-binding cassette, subfamily B (MDR/TAP), member 11; ATP-binding cassette, sub-family B (MDR/TAP), member 11; ATP-binding cassette, sub-family B, member 11; bile salt export pump; BRIC2; Bsep; Lith1; PFIC2; PFIC-2; PGY4; progressive familial intrahepatic cholestasis 2; sister of P-glycoprotein; sister p-glycoprotein; Spgp | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27413, 83569, 8647 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70127, O95342, Q9QY30 | |
| ABCB11 | |
| A synthesized peptide derived from human ABCB11(Accession O95342), corresponding to amino acid residues N665-V715. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction