missing translation for 'onlineSavingsMsg'
Learn More

ABCB5 Antibody (CL13618) [Alexa Fluor« 350], Novus Biologicals Biologicals™

Product Code. 30532407 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532407 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30532407 Supplier Novus Biologicals Supplier No. NBP344328AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABCB5 Monoclonal antibody specifically detects ABCB5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCB5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13618
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of Q2M3G0, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ABC Transporters, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 340273
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.