Learn More
ABCB5 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98737MFV500
This item is not returnable.
View return policy
Description
ABCB5 Polyclonal antibody specifically detects ABCB5 in Human samples. It is validated for Western Blot
Specifications
| mFluor Violet 500 SE | |
| ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
| Antigen and protein A Affinity-purified | |
| Primary |
| Western Blot | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human ABCB5. | |
| RUO | |
| Purified |