missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCB8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5122301

 View more versions of this product

Product Code. 17793694

  • € 180.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ABCB8 Polyclonal Antibody for Western Blot, IHC (P)

This nuclear gene encodes a multi-pass membrane protein that is targeted to the mitochondrial inner membrane. The encoded protein is an ATP-dependent transporter that may mediate the passage of organic and inorganic molecules out of the mitochondria. Loss of function of the related gene in mouse results in a disruption of iron homeostasis between the mitochondria and cytosol. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABCB8
Polyclonal
Unconjugated
ABCB8
4833412N02Rik; AA409895; ABC transporter 8; Abcb8; ATP binding cassette subfamily B member 8; ATP-binding cassette sub-family B member 8, mitochondrial; ATP-binding cassette, subfamily B (MDR/TAP), member 8; ATP-binding cassette, sub-family B (MDR/TAP), member 8; ATP-binding cassette, sub-family B, member 8; EST328128; hypothetical protein LOC548340; MABC1; M-ABC1; mitochondrial ABC protein; mitochondrial ATP-binding cassette 1; mitochondrial potassium channel ATP-binding subunit; mitochondrial sulfonylurea-receptor; MITOSUR; zgc:113037
Rabbit
Antigen affinity chromatography, Protein A
RUO
11194
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q9NUT2
ABCB8
Recombinant protein ATP Binding Cassette Transporter B8. The antigen corresponds to amino acid range 472-735 of the target protein.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.