missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB9 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB300-865
This item is not returnable.
View return policy
Description
ABCB9 Polyclonal specifically detects ABCB9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| ABCB9 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ABC transporter 9 protein, ATP-binding cassette sub-family B member 9, ATP-binding cassette transporter 9, ATP-binding cassette, sub-family B (MDR/TAP), member 9, EC 3.6.3, EC 3.6.3.44, EST122234, hABCB9, KIAA1520, TAPL, TAP-like protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23457 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 3 - 5 ug/ml, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| Q9NP78 | |
| ABCB9 | |
| Peptide with sequence C-GHNEPVANGSHKA corresponding to C-Terminus according to NP_062570.1, NP_062571.1, NP_982269.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both human isoforms of this protein (as represented by NP_062570.1, NP_062571.1and NP_982269.1). Variants (NP_062571.1; NP_982269.1) encode the same isoform. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction