missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBAS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99373-50ul
This item is not returnable.
View return policy
Description
GBAS Polyclonal antibody specifically detects GBAS in Human samples. It is validated for Western Blot
Specifications
| GBAS | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| glioblastoma amplified sequence, Glioblastoma-amplified sequence, NIPSNAP2NipSnap2,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2, protein NipSnap homolog 2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GBAS. | |
| 50 μg | |
| Neuroscience | |
| 2631 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction