Learn More
ABCC10 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30697-1-AP-150UL
 View more versions of this product
Product Code. 18087255
  • € 410.00 / 150µL
Description
The ABCC10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ABCC10. This antibody recognizes human, mouse antigen. The ABCC10 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
ABCC10
400 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
ABCC10, MRP7, SIMRP7
150 μL
89845
Store at -20°C. Stable for one year after shipment.