Learn More
ABCC10 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30697-1-AP-150UL
Description
The ABCC10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ABCC10. This antibody recognizes human, mouse antigen. The ABCC10 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| ABCC10 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| ABCC10, MRP7, SIMRP7 | |
| 150 μL | |
| 89845 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |