missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCC5 Antibody (OTI2C8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71540FR
This item is not returnable.
View return policy
Description
ABCC5 Monoclonal specifically detects ABCC5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ABCC5 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| ABCC5 | |
| Full length recombinant protein of human ABCC5 (NP_001018881) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2C8 | |
| Western Blot, Immunohistochemistry | |
| ATP-binding cassette sub-family C member 5, ATP-binding cassette, sub-family C (CFTR/MRP), member 5, canalicular multispecific organic anion transporter C, EST277145, MOAT-CABC33, MRP5MOATC, multidrug resistance-associated protein 5, Multi-specific organic anion transporter C, pABC11, SMRPDKFZp686C1782 | |
| Mouse | |
| Affinity Purified | |
| ABC Transporters, Amino Acids Drugs and other small molecules, Lipid and Metabolism | |
| 10057.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction