missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCC5, Mouse anti-Human, Clone: 1B12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010057-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: CITERGTHEELMNLNGDYATIFNNLLLGETPPVEINSKKETSGSQKKSQDKGPKTGSVKKEKAVKPEEGQLVQLEEKGQGSVPWSVYSpecifications
| ABCC5 | |
| Monoclonal | |
| Unconjugated | |
| ATP-binding cassette, sub-family C (CFTR/MRP), member 5 | |
| ABC33/DKFZp686C1782/EST277145/MOAT-C/MOATC/MRP5/SMRP/pABC11 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1B12 | |
| In 1x PBS, pH 7.4 | |
| NM_005688 | |
| ABCC5 | |
| ABCC5 (NP_005679.2, 760 a.a. ∼ 846 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10057 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction