Learn More
ABCC5 Mouse anti-Human, Unconjugated, Clone: 2H10F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67955-1-IG-20UL
 View more versions of this product
Product Code. 17521466
  • € 145.00 / 20µL
Description
The ABCC5 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human ABCC5. This antibody recognizes human, rat antigen. The ABCC5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ABCC5
Monoclonal
Unconjugated
20 μL
10057
IgG1
Western Blot
2H10F11
Mouse
Primary
Human, Rat