missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcc9, Mouse anti-Mouse, R-PE, Clone: S319A-14, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB18469.100ug
This item is not returnable.
View return policy
Description
Sequence: SSIVDAGLVLVFSEGILVECDTGPNLLQHKNGLFSTLVMTNKSpecifications
| Abcc9 | |
| Monoclonal | |
| R-PE | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| AI414027/AI449286/SUR2A/SUR2B/Sur2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S319A-14 | |
| Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ATP-binding cassette, sub-family C (CFTR/MRP), member 9 | |
| Abcc9 | |
| Recombinant protein corresponding to amino acids 1505-1546 at C-terminus of mouse Abcc9. | |
| RUO | |
| 20928 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction