Learn More
ABCD1 Mouse anti-Human, Clone: OTI2C12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF803207
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ABCD1 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. This peroxisomal membrane protein is likely involved in the peroxisomal transport or catabolism of very long chain fatty acids. Defects in this gene have been identified as the underlying cause of adrenoleukodystrophy, an X-chromosome recessively inherited demyelinating disorder of the nervous system.Specifications
| ABCD1 | |
| Monoclonal | |
| Unconjugated | |
| ABCD1 | |
| ABC42, ALD, ALDP, AMN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 215 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P33897 | |
| ABCD1 | |
| Human recombit protein fragment corresponding to amino acids 508-745 of human ABCD1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.