missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582591
This item is not returnable.
View return policy
Description
Immunogen sequence: KKDVDDDGEE KELMERLKKL SVPTSDEEDE VPAPKPRGGK KTKGGNVFAA LIQDQSEEEE EEEKHPPKPA KPEKNRINKA VSEEQQPALK GKKGKEEKSK GKAKPQNKFA AL Highest antigen sequence indentity to the following orthologs: Rat - 72%, Mouse - 73%.
This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
Specifications
| ABCF1 | |
| Polyclonal | |
| Unconjugated | |
| Abcf1 | |
| ABC27; ABC50; ABCF1; ATP binding cassette subfamily F member 1; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette sub-family F member 1; LOW QUALITY PROTEIN: ATP-binding cassette sub-family F member 1; ATP-binding cassette, subfamily F (GCN20), member 1; ATP-binding cassette, sub-family F (GCN20), member 1; ATP-binding cassette, sub-family F, member 1; AU041969; D17Wsu166e; GCN20; TNF-alpha stimulated ABC protein; TNFalpha-inducible ATP-binding protein; TNF-alpha-stimulated ABC protein; wu:fb79c06; wu:fc39a05; wu:fj94a08; zgc:85667 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NE71 | |
| Abcf1 | |
| Recombinant protein corresponding to Human ABCF1. Recombinant protein control fragment (Product #RP-90669). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction