missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABCF1 Recombinant Rabbit Monoclonal Antibody (4P9V8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537931
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, HepG2 Immunogen sequence: GQKARVVFAE LACREPDVLI LDEPTNNLDI ESIDALGEAI NEYKGAVIVV SHDARLITET NCQLWVVEEQ SVSQIDGDFE DYKREVLEAL GEVMVSRPRE.
This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
Specifications
| ABCF1 | |
| Recombinant Monoclonal | |
| 0.68 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6MG08, Q8NE71 | |
| Abcf1 | |
| A synthetic peptide corresponding to a sequence within amino acids 746-845 of human ABCF1 (Q8NE71). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4P9V8 | |
| Unconjugated | |
| Abcf1 | |
| ABC27; ABC50; ABCF1; ATP binding cassette subfamily F member 1; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette sub-family F member 1; LOW QUALITY PROTEIN: ATP-binding cassette sub-family F member 1; ATP-binding cassette, subfamily F (GCN20), member 1; ATP-binding cassette, sub-family F (GCN20), member 1; ATP-binding cassette, sub-family F, member 1; AU041969; D17Wsu166e; GCN20; TNF-alpha stimulated ABC protein; TNFalpha-inducible ATP-binding protein; TNF-alpha-stimulated ABC protein; wu:fb79c06; wu:fc39a05; wu:fj94a08; zgc:85667 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23, 85493 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction