missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABCF1 Recombinant Rabbit Monoclonal Antibody (SN07-12)
GREENER_CHOICE

Product Code. 16395724
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16395724 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16395724 Supplier Invitrogen™ Supplier No. MA532313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone SN07-12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Abcf1
Gene Accession No. Q6MG08, Q8NE71
Gene Alias ABC27; ABC50; ABCF1; ATP binding cassette subfamily F member 1; ATP-binding cassette 50; ATP-binding cassette 50 (TNF-alpha stimulated); ATP-binding cassette sub-family F member 1; ATP-binding cassette sub-family F member 1; LOW QUALITY PROTEIN: ATP-binding cassette sub-family F member 1; ATP-binding cassette, subfamily F (GCN20), member 1; ATP-binding cassette, sub-family F (GCN20), member 1; ATP-binding cassette, sub-family F, member 1; AU041969; D17Wsu166e; GCN20; TNF-alpha stimulated ABC protein; TNFalpha-inducible ATP-binding protein; TNF-alpha-stimulated ABC protein; wu:fb79c06; wu:fc39a05; wu:fj94a08; zgc:85667
Gene Symbols Abcf1
Host Species Rabbit
Immunogen Synthetic peptide within Human ABCF1 aa 796-845.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23, 85493
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.