missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCF2 Monoclonal Antibody (OTI8F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811105
This item is not returnable.
View return policy
Description
ABCF2 is a member of the ABC Transporter superfamily and of the GCN20 subfamily. As a member of the ABC Transporter family, it functions to transport various molecules across extra- and intracellular membranes.Specifications
| ABCF2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99LE6, Q9UG63 | |
| ABCF2 | |
| Human recombit protein fragment corresponding to amino acids 1-235 of human ABCF2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8F4 | |
| Unconjugated | |
| ABCF2 | |
| ABC28, EST133090, HUSSY-18, HUSSY18 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10061, 27407 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction