missing translation for 'onlineSavingsMsg'
Learn More

ABCF2 Mouse anti-Human, Mouse, Clone: OTI8F4, lyophilized, TrueMAB™

Product Code. 17611562
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17611562 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17611562 Supplier Origene Technologies Supplier No. CF811105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

ABCF2 is a member of the ABC Transporter superfamily and of the GCN20 subfamily. As a member of the ABC Transporter family, it functions to transport various molecules across extra- and intracellular membranes.

Specifications

Antigen ABCF2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8F4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ABCF2
Gene Accession No. Q99LE6, Q9UG63
Gene Alias ABC28, EST133090, HUSSY-18, HUSSY18
Gene Symbols ABCF2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-235 of human ABCF2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10061, 27407
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.