Learn More
Invitrogen™ ABCG1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA572939
Product Code. 15931595
  • € 532.00 / 0.10mL
Description
ABCG1 Polyclonal Antibody for Western Blot, IHC (P), Flow
ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. The ABC transporter ABCG1 (ABC8) is expressed on the cell surface and in intracellular compartments of cholesterol-laden macrophages and is induced in monocyte-derived macrophages during cholesterol influx mediated by acetylated low-density lipoprotein. In contrast, lipid efflux in cholesterol-laden macrophages, mediated by the cholesterol acceptor HDL(3), suppresses the expression of ABCG1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ABCG1
Polyclonal
Unconjugated
ABCG1
ABC transporter 8; ABC8; ABCG1; ATP binding cassette subfamily G member 1; ATP-binding cassette 8; ATP-binding cassette sub-family G member 1; ATP-binding cassette transporter 8; ATP-binding cassette transporter member 1 of subfamily G; ATP-binding cassette, subfamily G (WHITE), member 1; ATP-binding cassette, sub-family G (WHITE), member 1; ATP-binding cassette, subfamily G, member 1; AW413978; homolog of Drosophila white; I79_011524; MGC34313; White; White protein homolog; white protein homolog (ATP-binding cassette transporter 8); WHITE1; WHT1
Rabbit
Antigen affinity chromatography
RUO
100759619, 103346773, 11307, 85264, 9619
4°C, do not freeze
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P45844, Q64343
ABCG1
A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400).
100 μL
Primary
Human, Mouse, Rat, Rabbit, Monkey, Hamster