Learn More
Invitrogen™ ABCG1 Polyclonal Antibody
Chicken Polyclonal Antibody
Brand: Invitrogen™ PA19535
Description
PA1-9535 detects ABCG1 in human samples. PA1-9535 has been successfully used in Western blot procedures. By Western blot, PA1-9535 detects a 53 kDa protein representing ABCG1. PA1-9535 immunogen is a purified recombinant ABCG1 corresponding to the chromosome 21 gene (Human).
ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. The ABC transporter ABCG1 (ABC8) is expressed on the cell surface and in intracellular compartments of cholesterol-laden macrophages and is induced in monocyte-derived macrophages during cholesterol influx mediated by acetylated low-density lipoprotein. In contrast, lipid efflux in cholesterol-laden macrophages, mediated by the cholesterol acceptor HDL(3), suppresses the expression of ABCG1.
Specifications
| ABCG1 | |
| Polyclonal | |
| Unconjugated | |
| ABCG1 | |
| ABC transporter 8; ABC8; ABCG1; ATP binding cassette subfamily G member 1; ATP-binding cassette 8; ATP-binding cassette sub-family G member 1; ATP-binding cassette transporter 8; ATP-binding cassette transporter member 1 of subfamily G; ATP-binding cassette, subfamily G (WHITE), member 1; ATP-binding cassette, sub-family G (WHITE), member 1; ATP-binding cassette, subfamily G, member 1; AW413978; homolog of Drosophila white; I79_011524; MGC34313; White; White protein homolog; white protein homolog (ATP-binding cassette transporter 8); WHITE1; WHT1 | |
| Chicken | |
| IgY fraction | |
| RUO | |
| 9619 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P45844 | |
| ABCG1 | |
| Purified recombinant ABCG1 corresponding to the chromosome 21 gene (Human). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.