Learn More
ABCG2/CD338 Antibody (3G8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-22124APC
Product Code. 18489871
  • ONLINE EXCLUSIVE PRICE
    € 577.00
    € 544.95 / 0.10mL
    Save € 32.05
    5% Off
Description
ABCG2/CD338 Monoclonal antibody specifically detects ABCG2/CD338 in Human, Mouse, Primate samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ABCG2/CD338
Monoclonal
APC
PBS
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human, Mouse, Primate
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
3G8
Flow Cytometry
ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Purified recombinant fragment of human ABCG2 expressed in E. coli. [Uniprot: Q9UNQ0]
0.1 mL
ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Stem Cell Markers, Stem Cells
9429
Store at 4C in the dark.