missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92641-0.1ml
This item is not returnable.
View return policy
Description
ABCG8 Polyclonal antibody specifically detects ABCG8 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ABCG8 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human ABCG8 (NP_071882.1). MAGKAAEERGLPKGATPQDTSGLQDRLFSSESDNSLYFTYSGQPNTLEVRDLNYQVDLASQVPWFEQLAQFKMPWTSPSCQNSCELGIQNLSFKVRSGQMLAIIGSSGCGRASLLDVITGRGHGGKIKSGQIWINGQPSSPQLVRKCVAHVRQHNQLLPNLTVRETLAFIAQMRLPRTFSQAQRDKRVEDVIAELRLRQCADTRVGNMYVRGLSGGERRRVSIGVQLLWNPGILILDEPTSGLDSFTAHNLVKTLSRLAKGNRLVLISLHQPRSDIFRLFDLVLLMTSGTPIYLGAAQHMVQYFTAIGYPCPRYSNPADF | |
| 0.1 mL | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 64241 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction