missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG8 Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB400-117JF635
This item is not returnable.
View return policy
Description
ABCG8 Polyclonal antibody specifically detects ABCG8 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| ABCG8 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 635 | |
| ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
| A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProt# Q9H221] | |
| 0.2 mL | |
| ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 64241 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction