Learn More
B Braun™ Abdominal Scissors
Versatile, multipurpose scissors used for both dissection and surgical applications, heavier than Mayo scissors.
Brand: B Braun™ BC774
This item is not returnable.
View return policy
Description
- Manufactured from stainless steel
- Meticulously crafted to have a solid feel and to perform like an extension of the surgeon’s own hand
- Manufactured to strict guidelines for precision and reliability
Specifications
Abdominal Scissors | |
180 mm | |
Multipurpose, dissection, surgical |
Straight | |
Stainless Steel | |
Blunt/Blunt |