Learn More
Thermo Scientific™ Abgene™ SmartCap Mats
Description
The Thermo Scientific™ Abgene™ SmartCap Mat seals 96 samples at one time in a high-density storage format,affording maximum space saving benefits. Suitable for use with Thermo Scientific storage tubes ranging from 0.3 ml to1.4 ml, the Abgene SmartCap Mat maintain sa consistent seal down to -80°C.
Features:
- Efficient Storage
- Enhanced Materials
- Ease of Installation and Access
Specifications
Specifications
| Description | Seals 96 sample tubes at one time |
| No. of Wells | 96 |
| Product Type | Sealing Mat |
| Temperature Range (Metric) | -80°C |