Learn More
Thermo Scientific™ VersiPlate Frame, 96-well, skirted, white
Product Code. 15235156
Change view
Click to view available options
Quantity:
25 plates
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15235156 25 plates 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 15235156 Supplier Thermo Scientific™ Supplier No. AB1805
Only null left
Add to basket
Add to basket
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Flexible 96-well plates of low profile strip tubes for use in PCR and qPCR applications.
The Thermo Scientific VersiPlates are unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
Highlights
  • Easier sample handling as multiple strips of samples can be handled simultaneously
  • Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
  • Can be used in instruments with or without a VersiFrame
  • The reusable polycarbonate frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
  • The low profile format of the VersiPlate has a lower dead volume which reduces sample evaporation
  • Available in white which improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
  • Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals
Applications
  • PCR
  • qPCR/Real-Time PCR
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Packaging 25 Plate
Product Line VersiPlate
Color White
Product Type Frame
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Material Polycarbonate
No. of Wells 96
Plate Compatibility 96-Well Plates
Quantity 25 plates
Reaction Speed Low profile