Learn More
ABH1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98658MFV450
Product Code. 18527841
  • € 428.00 / 0.10mL
Description
ABH1 Polyclonal antibody specifically detects ABH1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ABH1
ABH1, ABHalkB, alkylation repair homolog (E. coli), alkB, alkB, alkylation repair homolog 1 (E. coli), ALKBH, alkylated DNA repair protein alkB homolog 1, alkylation repair, alkB homolog, Alpha-ketoglutarate-dependent dioxygenase ABH1, DNA lyase ABH1, EC 1.14.11.-, EC 4.2.99.18, hABH
0.1 mL
Immunoprecipitation, Immunohistochemistry (Paraffin)
Antigen and protein A Affinity-purified
8846
Thank You! Votre commentaire a été soumis