missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABH1 Antibody (OTI4C4), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71390G
This item is not returnable.
View return policy
Description
ABH1 Monoclonal specifically detects ABH1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| ABH1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ALKBH1 | |
| Recombinant protein fragment corresponding to amino acids 1-245 of human ABH1 (NP_006011) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| OTI4C4 | |
| Immunohistochemistry | |
| ABH1, ABHalkB, alkylation repair homolog (E. coli), alkB, alkB, alkylation repair homolog 1 (E. coli), ALKBH, alkylated DNA repair protein alkB homolog 1, alkylation repair, alkB homolog, Alpha-ketoglutarate-dependent dioxygenase ABH1, DNA lyase ABH1, EC 1.14.11.-, EC 4.2.99.18, hABH | |
| Mouse | |
| Affinity Purified | |
| DNA Repair | |
| 8846.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction