missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99129-100ul
This item is not returnable.
View return policy
Description
ABHD11 Polyclonal antibody specifically detects ABHD11 in Human samples. It is validated for Western Blot
Specifications
| ABHD11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| abhydrolase domain containing 11, abhydrolase domain-containing protein 11, EC 3.-, PP1226, WBSCR21, Williams Beuren syndrome chromosome region 21, Williams-Beuren syndrome chromosomal region 21 protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ABHD11. | |
| 100 μg | |
| Lipid and Metabolism | |
| 83451 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction