missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Negative Control for Rabbit Monoclonal Antibody (NCRBM/1520R), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-54471MFV450
This item is not returnable.
View return policy
Description
Negative Control for Rabbit Monoclonal Monoclonal antibody specifically detects Negative Control for Rabbit Monoclonal in Non-species specific samples. It is validated for Flow Cytometry
Specifications
| Negative Control for Rabbit Monoclonal | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Recombinant full-length human Negative Control for Rabbit Monoclonal protein | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG κ |
| Flow Cytometry | |
| NCRBM/1520R | |
| Flow Cytometry | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction